Parker Hannifin Compression Fitting Ferrules
Proveedor: Parker Hannifin
MFLX03302-26EA 9 EUR
MFLX03302-26 MFLX03302-36 MFLX03302-41 MFLX03302-31
Parker Hannifin Compression Fitting Ferrules
Racores y conectores
Get a secure seal at higher pressures.
  • Constructed of durable stainless steel
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Ferrules are required to create a leak-tight seal.