Flux® Pumps Air-Powered Drum Pump Motors
Proveedor: Flux Pumps
MFLX07070-70EA 1260 EUR
MFLX07070-70
Flux® Pumps Air-Powered Drum Pump Motors
Bombas Bombas para bidón
Configurable to your requirements.
  • High-viscosity tube set handles fluids with viscosities to 50,000 cp!
Air-operated motors are ideal for hazardous duty applications or where electricity is not available. Models 07070-60 and -70 are fitted with a muffler and include a grounding post. Model 07070-70 also has a unique trigger valve handle—press switch for intermittent pumping; lock switch for continuous use.
Models 70603-50, -52, and -54 feature variable speed control up to 900 rpm. These cool-running motors will not burn out. They are compact, portable, and operate in any position—ideal for transferring high-viscosity fluids.