Manual rotary microtome, RM 2235
Proveedor: Leica
720-2265EA 181 EUR
720-2265 720-1773
Manual rotary microtome, RM 2235
Microtomas Micrótomos
The manual rotary microtome for outstanding durability, precision, and comfort.
  • Universal cassette clamp for single-hand operation – Easy way to clamp/remove specimens
  • Optimised microtome base plate provides maximum rigidity
  • One-piece housing made from specially resistant plastic, without any edges where debris could accumulate, which reduces cleaning
  • Fewer clamping levers make the RM2235 easy to use by providing improved access to the cutting area
The RM2235 is the instrument of choice for all clinical histology and histopathology institutions.