Esta buscando: Anticuerpos
<B>IgGy Antibody Selector</B> – Quickly search hundreds of thousands of antibodies available for purchase from VWR by selecting common antibody features like antigen symbol and name, reactivity, clonality, conjugation, host, and other key factors. Antibodies used to identify and locate intracellular and extracellular proteins in common applications such as Western Blot, ELISA, ImmunoChemistry and Flow Cytometry are all available for your research.
IgGy Antibody Selector – Quickly search hundreds of thousands of antibodies available for purchase from VWR by selecting common antibody features like antigen symbol and name, reactivity, clonality, conjugation, host, and other key factors. Antibodies used to identify and locate intracellular and extracellular proteins in common applications such as Western Blot, ELISA, ImmunoChemistry and Flow Cytometry are all available for your research.
478 633  results were found
SearchResultCount:"478633"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Numero del catalogo: (ALEXENZABS2190100)
Proveedor: Enzo Life Sciences
Descripción: Recommended Applications: ELISA, ICC, IHC, WB
Species reactivity: Mammalian
UOM: 1 * 100 µl
Proveedor: US Biological
Descripción: HEAT SHOCK PROTEIN 27, PHOSPHO PAB RB XH 1 * 100 µl
Numero del catalogo: (USBI034772-HRP)
Proveedor: US Biological
Descripción: Anti-DPEP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
UOM: 1 * 200 µl
Numero del catalogo: (BOSSBS-0272R)
Proveedor: Bioss
Descripción: Catalyzes the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, an intermediate in the biosynth...
UOM: 1 * 100 µl
Numero del catalogo: (BOSSBS-11958R)
Proveedor: Bioss
Descripción: Transcription factors, OTX1 and OTX2, are two murine homologs of the Drosophila orthodenticle (OTD), show a limited amino acid sequence divergence. OT...
UOM: 1 * 100 µl
Numero del catalogo: (BOSSBS-11253R)
Proveedor: Bioss
Descripción: Anti-PIST Rabbit Polyclonal Antibody
UOM: 1 * 100 µl
Numero del catalogo: (AGRIAS101138)
Proveedor: Agrisera
Descripción: Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
UOM: 1 * 2 mg
Numero del catalogo: (BOSSBS-12540R)
Proveedor: Bioss
Descripción: ATF2 is a member of the ATF/CREB family of basic region leucine zipper DNA binding proteins that regulates transcription by binding to a consensus cAM...
UOM: 1 * 100 µl
Numero del catalogo: (AGRIAS101201)
Proveedor: Agrisera
Descripción: Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 488)
UOM: 1 * 1 mg
Numero del catalogo: (BOSSBS-2368R)
Proveedor: Bioss
Descripción: Anti-MAPT Rabbit Polyclonal Antibody
UOM: 1 * 100 µl
Proveedor: Tonbo Biosciences
Descripción: The L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset o...
Numero del catalogo: (BOSSBS-6100R)
Proveedor: Bioss
Descripción: No data available.
UOM: 1 * 100 µl
Numero del catalogo: (BOSSBS-9538R)
Proveedor: Bioss
Descripción: Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of ...
UOM: 1 * 100 µl
Numero del catalogo: (BOSSBS-4901R)
Proveedor: Bioss
Descripción: The Annexins are a family of structurally similar proteins. Annexins bind to phospholipids and may be involved in regulation of membrane transport, me...
UOM: 1 * 100 µl
Numero del catalogo: (BOSSBS-5941R)
Proveedor: Bioss
Descripción: RRAS2 (Related RAS viral oncogene homolog 2) is a member of the Ras superfamily. It is a GTP binding protein with GTPase activity. It might transduce ...
UOM: 1 * 100 µl
Numero del catalogo: (BOSSBS-9069R)
Proveedor: Bioss
Descripción: Anti-HN1 Rabbit Polyclonal Antibody
UOM: 1 * 100 µl
Consulta de precio
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta. Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta.
Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
El producto(s) marcados con este símbolo están descatalogados - se venden hasta acabar stock. Pueden encontrar alternativas buscando con el número de catálogo VWR listado arriba. Si necesita más ayuda, por favor llame al Servicio de atención al cliente de VWR al 902.222.897.
1 - 16 of 478 633
no targeter for Bottom