pCMV-script vectors
Proveedor: Agilent Technologies
AGLS212220EA 565 EUR
AGLS212220 AGLS212224
pCMV-script vectors
Vectores, plásmidos e bibliotecas
pCMV-Script PCR cloning system is a polymerase chain reaction (PCR) cloning method that can be performed in 1-hour without adding bases to the primers. The kit permits the efficient cloning of PCR fragments with a high yield and a low rate of false positives.
  • CMV promoter for high-level protein expression in mammalian cells
  • Large multiple cloning site contains 15 unique sites
  • Clone PCR products generated with any PCR enzyme
Precaución: For Research Use Only. Not for Use in Diagnostic Procedures.