G148 disulfato 50 mg/ml en solución acuosa, estéril
Sinónimos: Geneticin®
AGLS200049EA 122 EUR
AGLS200049
G148 disulfato 50 mg/ml en solución acuosa, estéril
Disulfato G148
G418 blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells.
  • Selects eukaryotic cells stably transfected with neomycin-resistant genes
  • Useful as a selective agent in gene transfer experiments
  • Supplied as a crystalline solid or aqueous solution
Precaución: For research use only. Not for use in diagnostic procedures.
Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692,72 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00058314
Núm. CAS: 108321-42-2