T4 DNA ligase
Proveedor: Agilent Technologies
AGLS600011EA 260 EUR
AGLS600011
T4 DNA ligase
Reactivos de ácidos nucleicos Nucleic Acid Modification and Labelling
T4 DNA ligase is used to ligate cohesive ends in an overnight reaction.
  • High efficiency at joining fragments with complementary sticky ends
It catalyses the linkage of adjacent 5'-phosphate and 3'-hydroxy ends of double-stranded DNA by formation of a phosphodiester bond.
Precaución: For research use only. Not for use in diagnostic procedures.