Thorium, plasma standard, BAKER INSTRA-ANALYZED® reagent
BAKR5750.0100EA 173 EUR
BAKR5750.0100
Thorium, plasma standard, BAKER INSTRA-ANALYZED® reagent
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022