Ethyl-6-fluoroimidazo[1,2-a]pyridine-3-carboxylate
Proveedor: Apollo Scientific
APOSPC510168-1GEA
80
EUR
APOSPC510168-1G
Ethyl-6-fluoroimidazo[1,2-a]pyridine-3-carboxylate
Ethyl-6-fluoroimidazo[1,2-a]pyridine-3-carboxylate
Formula:
C₁₀H₉FN₂O₂ MW: 208,1900 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD21364586 Núm. CAS: 1359655-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...