2,3,5-Trifluoro-4-methylbenzoic acid
Proveedor: Apollo Scientific
APOSPC448033-1GEA
326
EUR
APOSPC448033-1G
2,3,5-Trifluoro-4-methylbenzoic acid
2,3,5-Trifluoro-4-methylbenzoic acid
Formula:
C₈H₅F₃O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00456800 Núm. CAS: 773869-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...