2,6-Dichloro-3-(trifluoromethyl)benzyl alcohol 98%
Proveedor: Apollo Scientific
APOSPC302907-5GEA
184
EUR
APOSPC302907-5G
2,6-Dichloro-3-(trifluoromethyl)benzyl alcohol 98%
2,6-Dichloro-3-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₅Cl₂F₃O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11519348 Núm. CAS: 1092461-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...