4-(3-Aminopropoxy)benzonitrile 95%
Proveedor: Apollo Scientific
APOSOR965464-250MGEA
219
EUR
APOSOR965464-250MG
APOSOR965464-1G
4-(3-Aminopropoxy)benzonitrile 95%
4-(3-Aminopropoxy)benzonitrile
Formula:
C₁₀H₁₂N₂O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD16495890 Núm. CAS: 116753-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...