2',5'-Dimethoxyacetanilide 95%
Proveedor: Apollo Scientific
APOSOR950209-25GEA
132
EUR
APOSOR950209-25G
2',5'-Dimethoxyacetanilide 95%
2',5'-Dimethoxyacetanilide
Formula:
C₁₀H₁₃NO₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00043826 Núm. CAS: 3467-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...