3-(2-Bromophenyl)-5-(2-chlorophenyl)-1,2,4-oxadiazole
Proveedor: Apollo Scientific
APOSOR59381-1GEA
176
EUR
APOSOR59381-1G
3-(2-Bromophenyl)-5-(2-chlorophenyl)-1,2,4-oxadiazole
3-(2-Bromophenyl)-5-(2-chlorophenyl)-1,2,4-oxadiazole
Formula:
C₁₄H₈BrClN₂O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02217449 Núm. CAS: 1000339-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...