3-Carboxy-2-methoxyphenylboronic acid 98%
APOSOR3782-1GEA
0
EUR
APOSOR3782-1G
3-Carboxy-2-methoxyphenylboronic acid 98%
3-Carboxy-2-methoxyphenylboronic acid
Formula:
C₈H₉BO₅ MW: 195,97 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD09027237 Núm. CAS: 913836-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...