3-tert-Butylphenylboronic acid 95+%
Proveedor: Apollo Scientific
APOSOR361622-5GEA
91.6
EUR
APOSOR361622-5G
3-tert-Butylphenylboronic acid 95+%
3-tert-Butylphenylboronic acid
Formula:
C₁₀H₁₅BO₂ MW: 178,0400 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11110647 Núm. CAS: 560132-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...