2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester 95+%
APOSOR360089-1GEA
0
EUR
APOSOR360089-1G
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester 95+%
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester
Formula:
C₁₅H₂₂BNO₃ MW: 275,16 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07781184 Núm. CAS: 947191-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...