trans-1,4-Cyclohexanedicarboxylic acid
Proveedor: Apollo Scientific
APOSOR350368-100GEA
75.7
EUR
APOSOR350368-100G
trans-1,4-Cyclohexanedicarboxylic acid
trans-1,4-Cyclohexanedicarboxylic acid
Formula:
C₈H₁₂O₄ MW: 172,18 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00066203 Núm. CAS: 619-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...