N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine 98%
Proveedor: Apollo Scientific
APOSOR345028-100GEA
88.7
EUR
APOSOR345028-100G
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine 98%
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine
Formula:
C₁₆H₁₅N₅ MW: 277,3200 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09028125 Núm. CAS: 152460-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...