2-Cyclohexylideneacetic acid 95%
Proveedor: Apollo Scientific
APOSOR32888-1GEA
161
EUR
APOSOR32888-1G
2-Cyclohexylideneacetic acid 95%
2-Cyclohexylideneacetic acid
Formula:
C₈H₁₂O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01815140 Núm. CAS: 1552-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...