8-Quinolineboronic acid pinacol ester
Proveedor: Apollo Scientific
APOSOR303463-5GEA
390
EUR
APOSOR303463-5G
APOSOR303463-500MG
APOSOR303463-1G
8-Quinolineboronic acid pinacol ester
8-Quinolineboronic acid pinacol ester
Formula:
C₁₅H₁₈BNO₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD05663894 Núm. CAS: 190788-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...