4-Chloroisoquinoline
APOSOR300106-500MGEA
0
EUR
APOSOR300106-500MG
4-Chloroisoquinoline
4-Chloroisoquinoline
Formula:
C₉H₆ClN MW: 163,6036 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00234490 Núm. CAS: 1532-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...