4-Benzyloxyindole-3-carbaldehyde 98%
APOSOR1672-5GEA
0
EUR
APOSOR1672-5G
APOSOR1672-10G
4-Benzyloxyindole-3-carbaldehyde 98%
4-Benzyloxyindole-3-carbaldehyde
Formula:
C₁₆H₁₃NO₂ MW: 251,28 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00152002 Núm. CAS: 7042-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...