4-(Chlorosulphonyl)benzoic acid 95+%
Proveedor: Apollo Scientific
APOSOR12901-25GEA
170
EUR
APOSOR12901-25G
4-(Chlorosulphonyl)benzoic acid 95+%
4-(Chlorosulphonyl)benzoic acid
Formula:
C₇H₅ClO₄S MW: 220,63 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00007448 Núm. CAS: 10130-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...