3-Chloro-6-(pyrrol-1-yl)pyridazine 98%
APOSOR11559-1GEA
0
EUR
APOSOR11559-1G
APOSOR11559-5G
3-Chloro-6-(pyrrol-1-yl)pyridazine 98%
3-Chloro-6-(pyrrol-1-yl)pyridazine
Formula:
C₈H₆ClN₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00828665 Núm. CAS: 5096-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...