LOXO-195 98%

Proveedor: Apollo Scientific

APOSBISN0067-5MGEA 154 EUR
APOSBISN0067-5MG APOSBISN0067-10MG
LOXO-195 98%
LOXO-195

LOXO-195 is a new-generation and selective TRK inhibitor.

Formula: C₂₀H₂₁FN₆O
MW: 380,43 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD31620755
Núm. CAS: 2097002-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR