6-Ethoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
Proveedor: Apollo Scientific
|
Atención
|
APOSOR31010-250MGEA
21.9
EUR
APOSOR31010-250MG
APOSOR31010-1G
6-Ethoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
6-Ethoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
Formula:
C₁₄H₁₈N₂O₃·ClH MW: 298,765 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD29042693 Núm. CAS: 1823188-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...