4,8-Dibromoquinoline-3-carbonitrile
APOSOR309347-1GEA
0
EUR
APOSOR309347-1G
4,8-Dibromoquinoline-3-carbonitrile
4,8-Dibromoquinoline-3-carbonitrile
Formula:
C₁₀H₄Br₂N₂ MW: 311,96 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD15527484 Núm. CAS: 1242260-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...