6-Bromo-2-hydrazino-3-methylquinoline hydrochloride

Proveedor: Apollo Scientific

APOSOR307524-1GEA 384 EUR
APOSOR307524-1G
6-Bromo-2-hydrazino-3-methylquinoline hydrochloride
6-Bromo-2-hydrazino-3-methylquinoline hydrochloride
Formula: C₁₀H₁₀BrN₃·ClH
MW: 288,571 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11505170
Núm. CAS: 1172069-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR