6-Iodo-[1,2,4]triazolo[1,5-a]pyridine 97%
APOSOR307049-100MGEA
0
EUR
APOSOR307049-100MG
APOSOR307049-250MG
6-Iodo-[1,2,4]triazolo[1,5-a]pyridine 97%
6-Iodo-[1,2,4]triazolo[1,5-a]pyridine
Formula:
C₆H₄IN₃ MW: 245,021 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06797552 Núm. CAS: 614750-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...