2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride
Proveedor: Apollo Scientific
|
Atención
|
APOSOR30669-100MGEA
94.5
EUR
APOSOR30669-100MG
2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride
2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride
Formula:
C₈H₁₃N₃O·ClH MW: 203,669 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11869758 Núm. CAS: 1214028-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...