(S)-tert-Butyl 3-(oxetan-3-ylamino)pyrrolidine-1-carboxylate

APOSOR306061-1GEA 0 EUR
APOSOR306061-1G
(S)-tert-Butyl 3-(oxetan-3-ylamino)pyrrolidine-1-carboxylate
(S)-tert-Butyl 3-(oxetan-3-ylamino)pyrrolidine-1-carboxylate
Formula: C₁₂H₂₂N₂O₃
MW: 242,315 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD20233198
Núm. CAS: 1349699-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR