4'-Bromobiphenyl-4-yl butyl ether
Proveedor: Apollo Scientific
APOSOR305566-1GEA
326
EUR
APOSOR305566-1G
4'-Bromobiphenyl-4-yl butyl ether
4'-Bromobiphenyl-4-yl butyl ether
Formula:
C₁₆H₁₇BrO MW: 305,21 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD26142691 Núm. CAS: 63619-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...