Methyl 3-(4-hydroxy-2-methoxyphenyl)propanoate
APOSOR305448-250MGEA
0
EUR
APOSOR305448-250MG
Methyl 3-(4-hydroxy-2-methoxyphenyl)propanoate
Methyl 3-(4-hydroxy-2-methoxyphenyl)propanoate
Formula:
C₁₁H₁₄O₄ MW: 210,226 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD24728371 Núm. CAS: 628333-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...