Methyl 6-chloro-1H-indazole-4-carboxylate
Proveedor: Apollo Scientific
APOSOR305441-250MGEA
242
EUR
APOSOR305441-250MG
Methyl 6-chloro-1H-indazole-4-carboxylate
Methyl 6-chloro-1H-indazole-4-carboxylate
Formula:
C₉H₇ClN₂O₂ MW: 210,617 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07781390 Núm. CAS: 885519-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...