1-[4-(1H-Pyrazolo[3,4-b]pyridin-4-yl)morpholin-2-yl]methanamine
Proveedor: Apollo Scientific
APOSOR305395-250MGEA
749
EUR
APOSOR305395-250MG
1-[4-(1H-Pyrazolo[3,4-b]pyridin-4-yl)morpholin-2-yl]methanamine
1-[4-(1H-Pyrazolo[3,4-b]pyridin-4-yl)morpholin-2-yl]methanamine
Formula:
C₁₁H₁₅N₅O MW: 233,27 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD13183749 Núm. CAS: 1034769-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...