4-Methoxy-1,2-dihydro-3H-indazol-3-one
APOSOR305091-250MGEA
0
EUR
APOSOR305091-250MG
4-Methoxy-1,2-dihydro-3H-indazol-3-one
4-Methoxy-1,2-dihydro-3H-indazol-3-one
Formula:
C₈H₈N₂O₂ MW: 164,161 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD13176928 Núm. CAS: 1000342-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...