tert-Butyl 3-bromo-5-cyano-1H-indazole-1-carboxylate
APOSOR305065-1GEA
0
EUR
APOSOR305065-1G
APOSOR305065-250MG
tert-Butyl 3-bromo-5-cyano-1H-indazole-1-carboxylate
tert-Butyl 3-bromo-5-cyano-1H-indazole-1-carboxylate
Formula:
C₁₃H₁₂BrN₃O₂ MW: 322,157 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD13183024 Núm. CAS: 473416-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...