7-Oxa-1-azaspiro[3.5]nonane
Proveedor: Apollo Scientific
APOSOR304425-1GEA
1560
EUR
APOSOR304425-1G
7-Oxa-1-azaspiro[3.5]nonane
7-Oxa-1-azaspiro[3.5]nonane
Formula:
C₇H₁₃NO MW: 127,184 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD14581251 Núm. CAS: 106404-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...