7-Oxospiro[3.5]nonane-2-carboxylic acid
Proveedor: Apollo Scientific
APOSOR304417-1GEA
1830
EUR
APOSOR304417-1G
7-Oxospiro[3.5]nonane-2-carboxylic acid
7-Oxospiro[3.5]nonane-2-carboxylic acid
Formula:
C₁₀H₁₄O₃ MW: 182,216 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD24857339 Núm. CAS: 1440962-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...