6-(2,6-Dimethylmorpholino)-2-methylpyrimidin-4-amine
APOSOR302557-1GEA
0
EUR
APOSOR302557-1G
6-(2,6-Dimethylmorpholino)-2-methylpyrimidin-4-amine
6-(2,6-Dimethylmorpholino)-2-methylpyrimidin-4-amine
Formula:
C₁₁H₁₈N₄O MW: 222,287 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09880707 Núm. CAS: 1023818-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...