5-Bromo-N-(oxetan-3-yl)pyrimidin-2-amine
Proveedor: Apollo Scientific
APOSOR302481-1GEA
1190
EUR
APOSOR302481-1G
5-Bromo-N-(oxetan-3-yl)pyrimidin-2-amine
5-Bromo-N-(oxetan-3-yl)pyrimidin-2-amine
Formula:
C₇H₈BrN₃O MW: 230,062 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD22690829 Núm. CAS: 1439823-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...