6-Ethyl-2-methylpyrimidin-4-amine
Proveedor: Apollo Scientific
APOSOR302376-1GEA
614
EUR
APOSOR302376-1G
6-Ethyl-2-methylpyrimidin-4-amine
6-Ethyl-2-methylpyrimidin-4-amine
Formula:
C₇H₁₁N₃ MW: 137,182 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09909802 Núm. CAS: 90008-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...