2,2-Dimethylpiperidine-3-carbonitrile hydrochloride

APOSOR302338-1GEA 0 EUR
APOSOR302338-1G
2,2-Dimethylpiperidine-3-carbonitrile hydrochloride
2,2-Dimethylpiperidine-3-carbonitrile hydrochloride
Formula: C₈H₁₄N₂·ClH
MW: 174,671 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD24368555
Núm. CAS: 1374658-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR