Ethyl 1-(5-(methylamino)pyridin-2-yl)-1H-pyrazole-4-carboxylate

APOSOR302179-1GEA 0 EUR
APOSOR302179-1G
Ethyl 1-(5-(methylamino)pyridin-2-yl)-1H-pyrazole-4-carboxylate
Ethyl 1-(5-(methylamino)pyridin-2-yl)-1H-pyrazole-4-carboxylate
Formula: C₁₂H₁₄N₄O₂
MW: 246,265 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD26403991
Núm. CAS: 1624261-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR