Methyl 4-acetyl-5-methylisoxazole-3-carboxylate
Proveedor: Apollo Scientific
APOSOR29702-250MGEA
219
EUR
APOSOR29702-250MG
APOSOR29702-1G
Methyl 4-acetyl-5-methylisoxazole-3-carboxylate
Methyl 4-acetyl-5-methylisoxazole-3-carboxylate
Formula:
C₈H₉NO₄ MW: 183,161 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00068116 Núm. CAS: 104149-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...