Ethyl 4-hydroxy-8-iodoquinoline-3-carboxylate ≥95%
Proveedor: Apollo Scientific
APOSOR183496-250MGEA
432
EUR
APOSOR183496-250MG
Ethyl 4-hydroxy-8-iodoquinoline-3-carboxylate ≥95%
Ethyl 4-hydroxy-8-iodoquinoline-3-carboxylate
Formula:
C₁₂H₁₀INO₃ MW: 343,117 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD06808574 Núm. CAS: 49713-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...