7-Bromo-4-hydroxyquinoline-3-carboxylic acid
Proveedor: Apollo Scientific
APOSOR183473-1GEA
74.2
EUR
APOSOR183473-1G
7-Bromo-4-hydroxyquinoline-3-carboxylic acid
7-Bromo-4-hydroxyquinoline-3-carboxylic acid
Formula:
C₁₀H₆BrNO₃ MW: 268,064 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02684222 Núm. CAS: 860205-92-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...