N-[3-(1H-Pyrazol-3-yl)phenyl]acetamide
Proveedor: Apollo Scientific
APOSOR183454-1GEA
183
EUR
APOSOR183454-1G
N-[3-(1H-Pyrazol-3-yl)phenyl]acetamide
N-[3-(1H-Pyrazol-3-yl)phenyl]acetamide
Formula:
C₁₁H₁₁N₃O MW: 201,225 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09028477 Núm. CAS: 197093-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...