(S)-1-Benzyl-3-isopropylpiperazine
APOSOR15646-1GEA
0
EUR
APOSOR15646-1G
(S)-1-Benzyl-3-isopropylpiperazine
(S)-1-Benzyl-3-isopropylpiperazine
Formula:
C₁₄H₂₂N₂ MW: 218,338 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03787917 Núm. CAS: 324748-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...